Learn More
Med Vet International McKesson Test Tube, Round Bottom, Plain, 13 X 75 mm, 5 mL, Without Color Coding, 177-110471, 1000/Bag
Catalog No. 503044180
Encompass_Preferred
McKesson Test Tube Round Bottom Plain 13 X 75 mm 5 mL Without Color Coding Without Closure Polypropylene Tube
Catalog No. 50-304-4180 Supplier Med Vet International Supplier No. 1175264BG
McKesson Test Tube Round Bottom Plain 13 X 75 mm 5 mL Without Color Coding Without Closure Polypropylene Tube