Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ TB Quat Disinfectant Ready-To-Use Cleaner

Catalog No. 17623920 Shop All 3M Products
Change view
Click to view available options
Volume (Metric):
0.946 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
17-623-920 0.946 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-623-920 Supplier 3M™ Supplier No. 7100034339
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ TB Quat Disinfectant Ready-To-Use Cleaner is a deodorizer, disinfectant, virucide, tuberculocide, mildewstat, and fungicide. This product contains 2100 ppm active germicide, making it effective against a broad spectrum of pathogenic microorganisms.

  • Kills SARS-CoV-2, which causes COVID-19 (SARS-Related Coronavirus 2) on hard non-porous surfaces in just 60 sec.
  • 5 min. contact time for Mycobacterium tuberculosis BCG (Mycobacterium bovis), Hepatitis B, and Hepatitis C
  • Ready-To-Use cleaner, deodorizer, disinfectant, virucide, tuberculocidal, mildewstat, and fungicide. Clean, deodorize and disinfect in one step when used according to disinfection directions.
  • No rinsing is necessary.
  • Contains 2100 ppm active germicide making it highly effective against a broad spectrum of pathogenic microorganisms (bacteria, antibiotic-resistant bacteria, viruses, fungi, mold, & mildew).
  • Non-Acid, Non-Abrasive: Designed specifically as a general, non-acid, non-abrasive cleaner and disinfectant for use in hospitals, nursing homes, hotels, schools, and other commercial/industrial buildings.
  • For Non-Porous Surfaces: Disinfects hard, non-porous, inanimate environmental surfaces like floors, walls, metal and stainless steel surfaces, glazed porcelain, glazed ceramic tile, plastic surfaces.
  • OSHA Bloodborne Pathogen Standard: Complies with standard for disinfecting surfaces soiled with blood or other potentially infectious body fluids.
  • Meets OSHA Bloodborne Pathogen Standard for HIV, HBV & HCV
TRUSTED_SUSTAINABILITY

Specifications

Product Type Disinfectant Cleaner
Form Liquid
Container Type Flip-top Bottle, Spray Bottle
Dilution Ready to use. Do not dilute.
Quantity 12/Cs.
Volume (English) 1 qt.
Volume (Metric) 0.946 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.