Learn More
StatLab TB Splash Proof Container
SDP
Catalog No. 21010248
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
21-010-248 Non-sterile
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-248 Supplier StatLab Supplier No. 252997
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Tuberculosis (TB) Splash Proof Container is made per CDC Recommendations to discard contaminated liquid safely.
  • Containers and funnel assembly is autoclavable and reusable
  • No rack
  • Set includes two 1 L containers and two funnels
Specifications
Product Type Disposable Sample Container
Closure Included Yes
Graduated No
Material Polypropylene
Shape Rectangular
Sterility Non-sterile