Learn More
Med Vet International McKesson Polystyrene Test Tube, 5 mL, 12 x 75 mm, 177-110410, 1000/Bag
Catalog No. 503042950
Encompass_Preferred
McKesson Test Tube Round Bottom Plain 12 X 75 mm 5 mL Without Color Coding Without Closure Polystyrene Tube 12 x 75 millimeters Molded from premium-grade polystyrene (PS) for clarity Can withstand 1500 RPM during centrifugation Tubes are nongraduated with round bottoms and no color coding Brittleness temperature: 20° C Maximum temperature: 90° C There are 1,000 5-mL polystyrene translucent test tubes in a bag
Catalog No. 50-304-2950 Supplier Med Vet International Supplier No. 1082088BG
McKesson Test Tube Round Bottom Plain 12 X 75 mm 5 mL Without Color Coding Without Closure Polystyrene Tube 12 x 75 millimeters Molded from premium-grade polystyrene (PS) for clarity Can withstand 1500 RPM during centrifugation Tubes are nongraduated with round bottoms and no color coding Brittleness temperature: 20° C Maximum temperature: 90° C There are 1,000 5-mL polystyrene translucent test tubes in a bag