Learn More
Med Vet International McKesson Test Tube, 13 x 100 mm, 177-1510, 1000/Case
Catalog No. 503042884
Encompass_Preferred
McKesson Test Tube Round Bottom Plain 13 X 100 mm 7 mL Without Color Coding Without Closure Glass Tube 13 x 100 mm 7 mL Premium-quality, clear glass tubes provide excellent chemical resistance and durability Chemically inert tubes, made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from tubes made from soda lime glass Round bottoms and fire-polished rims Can be used for tissue culture and blood banks Rigidly controlled dimensional specifications for reliable, uniform results Produced in an ISO 9001:2008 Certified Factory Specially packaged neatly in heavy-duty boxes, shrink-wrapped in plastic film to keep them particle-free and prevent breakage during shipment
Catalog No. 50-304-2884 Supplier Med Vet International Supplier No. 1082086CS
McKesson Test Tube Round Bottom Plain 13 X 100 mm 7 mL Without Color Coding Without Closure Glass Tube 13 x 100 mm 7 mL Premium-quality, clear glass tubes provide excellent chemical resistance and durability Chemically inert tubes, made from superior quality borosilicate glass to reduce pH changes and contaminants that are potentially leached from tubes made from soda lime glass Round bottoms and fire-polished rims Can be used for tissue culture and blood banks Rigidly controlled dimensional specifications for reliable, uniform results Produced in an ISO 9001:2008 Certified Factory Specially packaged neatly in heavy-duty boxes, shrink-wrapped in plastic film to keep them particle-free and prevent breakage during shipment