Learn More
Med Vet International VACUETTE Venous Blood Collection Tube Sodium Citrate Additive 3 mL Pull Cap Polyethylene Terephthalate (PET) Tube, 454334, 50/Pack
Catalog No. 503043575
Encompass_Preferred
Coagulation tubes have a volume of 3 mL Nonscrew safety pull cap can be removed with a gentle pull Pull caps are light blue with black rings Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label 18-month shelf life from date of manufacture
Catalog No. 50-304-3575 Supplier Med Vet International Supplier No. 444524PK
Coagulation tubes have a volume of 3 mL Nonscrew safety pull cap can be removed with a gentle pull Pull caps are light blue with black rings Mixing ratio is one part citrate to nine parts blood when tube is filled appropriately as indicated by fill mark on label 18-month shelf life from date of manufacture