Learn More
Teknova TB1 Broth. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387544
Encompass
1.0000% Tryptone, 0.5000% Sodium Chloride,0.0001% Thiamine HCl
Catalog No. 50-238-7544 Supplier Teknova Supplier No. T04092PK
1.0000% Tryptone, 0.5000% Sodium Chloride,0.0001% Thiamine HCl