Learn More
TARGETMOL CHEMICALS INC TBA-7371 25MG
SDP
Catalog No. 502576970
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TBA-7371 (DprE1-IN-1) is a potent inhibitor of DprE1 and PDE6. purity: 99%
Catalog No. 50-257-6970 Supplier TARGETMOL CHEMICALS INC Supplier No. T322425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. TBA-7371 (DprE1-IN-1) is a potent inhibitor of DprE1 and PDE6. purity: 99%