Learn More
Medchemexpress LLC HY-111747 10mg Medchemexpress, TBAJ-587 CAS:2252316-16-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11174710MG
Encompass_Preferred
Medchemexpress, HY-111747 10mg TBAJ-587 CAS:2252316-16-6 TBAJ-587, a potent anti-tuberculosis agent, inhibits M.tb strain H37Rv growth with MIC90s of 0.006 and <0.02 µg/mL in MABA and LORA assay, respectively. TBAJ-587 inhibits hERG channel minimally, attenuates inhibition of the cardiac potassium channel protein coded by the hERG, which is important for cardiac repolarization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8947