Learn More
TARGETMOL CHEMICALS INC TBAJ-587 5MG
SDP
Catalog No. 502880024
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. TBAJ-587 is a potent anti-tuberculosis agent. TBAJ-587 inhibits M.tb strain H37Rv growth with MIC90s of 0.006 and <0.02 ug/mL in MABA and LORA assay, respectively. BAJ-587 has more potent activity against M. tuberculosis and better efficacy in animal models of TB. Purity 98%
Catalog No. 50-288-0024 Supplier TARGETMOL CHEMICALS INC Supplier No. T59975MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. TBAJ-587 is a potent anti-tuberculosis agent. TBAJ-587 inhibits M.tb strain H37Rv growth with MIC90s of 0.006 and <0.02 ug/mL in MABA and LORA assay, respectively. BAJ-587 has more potent activity against M. tuberculosis and better efficacy in animal models of TB. Purity 98%