Learn More
Cell Biolabs Inc OxiSelect™ TBARS Assay Kit, 200 assays
Catalog No. NC9835067
Encompass_Preferred
Thiobarbituric Acid Reactive Substances (TBARS) is a well-established assay for screening and monitoring lipid peroxidation. The rapid and easy protocol has been modified by researchers in the evaluation of drugs, food, as well as human and animal tissue samples. MDA forms a 1:2 adduct with thiobarbituric acid. The MDA-TBA adduct formed from the reaction of MDA in samples with TBA can be measured colorimetrically or fluorometrically. TBARS levels are determined from a Malondialdehyde equivalence standard. The OxiSelect™ TBARS Assay Kit offers a simple, reproducible, and consistent system for the detection of lipid peroxidation in urine, plasma, serum, lysates, and tissue homogenates. This kit includes an MDA standard for use as a positive control. Each kit provides sufficient reagents to perform 200 tests including standard curve and unknown samples.
Catalog No. NC9835067 Supplier Cell Biolabs Inc Supplier No. STA330
Thiobarbituric Acid Reactive Substances (TBARS) is a well-established assay for screening and monitoring lipid peroxidation. The rapid and easy protocol has been modified by researchers in the evaluation of drugs, food, as well as human and animal tissue samples. MDA forms a 1:2 adduct with thiobarbituric acid. The MDA-TBA adduct formed from the reaction of MDA in samples with TBA can be measured colorimetrically or fluorometrically. TBARS levels are determined from a Malondialdehyde equivalence standard. The OxiSelect™ TBARS Assay Kit offers a simple, reproducible, and consistent system for the detection of lipid peroxidation in urine, plasma, serum, lysates, and tissue homogenates. This kit includes an MDA standard for use as a positive control. Each kit provides sufficient reagents to perform 200 tests including standard curve and unknown samples.