Learn More
Arbor Assays TBARS/MDA Universal Colorimetric Detection Kit, (2 Plate)
SDP
Catalog No. 502278518
Encompass_Preferred
The DetectX TBARS/MDA Universal Colorimetric Detection Kit allows assessment of lipid peroxidation without boiling water bath treatment.
This item is not returnable. View return policy
Catalog No. 50-227-8518 Supplier Arbor Assays Supplier No. K077H1
The DetectX TBARS/MDA Universal Colorimetric Detection Kit allows assessment of lipid peroxidation without boiling water bath treatment.