Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ TBARS Parameter Assay Kit

Catalog No. kge013
Change view
Click to view available options
Quantity:
2 x 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KGE013 2 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. KGE013 Supplier R&D Systems™ Supplier No. KGE013
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

TBARS Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The TBARS Parameter Assay Kit quantifies multi-species TBARS/Thiobarbituric Acid Reactive Substances in cell culture supernates (150 ul), cell lysates (150 ul), serum (150 ul), edta plasma (150 ul), heparin plasma (150 ul), urine (150 ul).

Specifications

Assay Range 0.00 - 16 uM
Assay Sensitivity 0.055 uM
Conjugate Biotin
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity TBARS in various samples
For Use With (Application) ELISA
Quantity 2 x 96-well Plates
Sample Volume Cell Culture Supernates (150uL),Cell Lysates (150uL),Serum (150uL),EDTA Plasma (150uL),Heparin Plasma (150uL),Urine (150uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.