Learn More
Selleck Chemical LLC TBB 25mg 17374-26-4 4,5,6,7-tetrabromobenzotriazole
SDP
Supplier:  Selleck Chemical LLC S526525MG
Encompass_Preferred
TBB (4,5,6,7-tetrabromobenzotriazole) is a selective cell-permeable CK2 inhibitor with IC50 values of 0.9 and 1.6 ?M for rat liver and human recombinant CK2 respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8333