Learn More
Abcam TBB, casein kinase-2 (CK2) inhibitor, 10MG
Catalog No. 502707056
Encompass_Preferred
MW 434.71 Da, Purity >98%. Selective, cell-permeable casein kinase-2 (CK2) inhibitor (IC₅₀ = 0.5 μM). Acts in an ATP/GTP competitive manner. Induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7056 Supplier Abcam Supplier No. AB12098810MG
MW 434.71 Da, Purity >98%. Selective, cell-permeable casein kinase-2 (CK2) inhibitor (IC₅₀ = 0.5 μM). Acts in an ATP/GTP competitive manner. Induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement