Learn More
Abcam TBB, casein kinase-2 (CK2) inhibitor, 50MG
Catalog No. 502707055
Encompass_Preferred
MW 434.71 Da, Purity >98%. Selective, cell-permeable casein kinase-2 (CK2) inhibitor (IC₅₀ = 0.5 μM). Acts in an ATP/GTP competitive manner. Induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7055 Supplier Abcam Supplier No. AB12098850MG
MW 434.71 Da, Purity >98%. Selective, cell-permeable casein kinase-2 (CK2) inhibitor (IC₅₀ = 0.5 μM). Acts in an ATP/GTP competitive manner. Induces apoptosis.
The product is subject to the following: Abcam Restricted Use Statement