Learn More
Cayman Chemical TBBPA monsulfate 10mg
SDP
Supplier:  Cayman Chemical 900456510 MG
Encompass_Preferred
A PPARγ agonist and active metabolite of TBBPA; increases PPARγ activity in a reporter assay using HGELN cells expressing PPARγ
Catalog No. 50-289-5325