Learn More
Aviva Systems Biology Corp TBC1D10A Antibody - C-terminal region (ARP60584_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60584_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D10A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TBC1D10A. Alternate names: EPI64, TBC1D10, dJ130H16.1, dJ130H16.2ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-287-091