Learn More
Aviva Systems Biology Corp TBC1D10B Antibody - C-terminal region (ARP34521_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34521_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D10B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TBC1D10B. Alternate names: EPI64B, FP2461ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55433