Learn More
Aviva Systems Biology Corp TBC1D10C Antibody - N-terminal region (ARP55893_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55893_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D10C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TBC1D10C. Alternate names: FLJ00332, MGC46488, EPI64C, CARABINApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-61652