Learn More
Aviva Systems Biology Corp TBC1D13 Antibody - middle region (ARP58536_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58536_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D13. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBC1D13. Alternate names: FLJ10743, RP11-545E17.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62348