Learn More
Aviva Systems Biology Corp TBC1D16 Antibody - middle region (ARP42534_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TBC1D16. Alternate names: FLJ20748, MGC25062ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-58257