Learn More
Aviva Systems Biology Corp TBC1D19 Antibody - middle region (ARP57195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D19. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TBC1D19. Alternate names: FLJ11082ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-274