Learn More
Aviva Systems Biology Corp TBC1D22A Antibody - N-terminal region (ARP55029_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D22A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TBC1D22A. Alternate names: C22orf4, HSC79E021ApplicationWB. Species reactivity: Human, Mouse, Pig, Rat
Catalog No. 50-014-61404