Learn More
Aviva Systems Biology Corp TBC1D25 Antibody - N-terminal region (ARP56369_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56369_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBC1D25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TBC1D25. Alternate names: MG81, MGC126866, MGC126868, MGC149731, MGC149732, OATL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61830