Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci TBC1D4 Peptide
SDP

Catalog No. 16917691
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-917-691 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-917-691 Supplier ProSci Supplier No. 4415P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

TBC1D4 peptide is used for blocking the activity of TBC1D4 antibody (catalog number 4415). It usually blocks the antibody activity completely in Western blot by incubating the peptide with equal volume of antibody for 30 min at 37°C.

Specifications

Gene ID (Entrez) 9882
Species Human
Accession Number NP_055647
Common Name TBC1 domain family, member 4
Concentration 200 μg/mL
Content And Storage Store TBC1D4 peptide at -20°C, stable for one year.
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol TBC1D4
Quantity 0.05 mg
Sequence 19 amino acids near the carboxy terminus of human TBC1D4.
Synonym AS160, AS160, KIAA0603, TBC1 domain family member 4, Akt substrate of 160 kDa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.