Learn More
Aviva Systems Biology Corp TBCCD1 Antibody - N-terminal region (ARP57123_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57123_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TBCCD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TBCCD1. Alternate names: FLJ10560ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit
Catalog No. 50-014-62085