Learn More
Med Vet International BD Vacutainer Plus Venous Blood Collection Tube Clot Activator / Separator Gel Additive 5 mL BD Hemogard™ Closure Plastic Tube, 367986, 1000/Case
Catalog No. 503044520
Encompass_Preferred
Not recommended for blood bank use BD Vacutainer brand sterile chemistry tubes with the BD Hemogard™ closure help to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact Additive: Clot activator and gel for serum separation Silicone coated interior Blood clotting time 30 minutes
Catalog No. 50-304-4520 Supplier Med Vet International Supplier No. 207068CS
Not recommended for blood bank use BD Vacutainer brand sterile chemistry tubes with the BD Hemogard™ closure help to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact Additive: Clot activator and gel for serum separation Silicone coated interior Blood clotting time 30 minutes