Learn More
Kd Medical Inc TBE 10X 1 LITER BOTTLE
SDP
Catalog No. 501018926
Encompass_Preferred
Tris Borate EDTA is used for polyacrylamide and agarose gel electrophoresis. 10X TBE is concentrated and ready to be diluted to a 1X working solution. One 1 liter bottle.
Catalog No. 50-101-8926 Supplier Kd Medical Inc Supplier No. RGF3330
Tris Borate EDTA is used for polyacrylamide and agarose gel electrophoresis. 10X TBE is concentrated and ready to be diluted to a 1X working solution. One 1 liter bottle.