Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBE 10X Buffer, MP Biomedicals™

Catalog No. MP1TBE10X02 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1TBE10X02 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1TBE10X02 Supplier MP Biomedicals, Inc Supplier No. 11TBE10X02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TBE 10X Buffer

  • TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis, after dilution to working concentrations.
  • TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.
  • TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.
  • TBE buffer maintains the structural integrity of nucleic acids and more suitable for their size analysis.
  • TBE has a greater buffering capacity and will give sharper resolution. TBE is better suited for high-voltage (>150 V) electrophoresis because of its higher buffering capacity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00236358
Synonym Tris-Borate-EDTA buffer

Specifications

Color Clear Colorless
CAS 7732-18-5
For Use With (Application) Electrophoresis
pH 8.2 to 8.4 (literature)
Physical Form Solution
Quantity 1 L
Solution Type Buffer
Solubility Information Soluble in water.
Grade Molecular Biology
storageNote1 Store at Room temperature(15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.