Learn More
Research Products International Corp TRIS-BORATE-EDTA, 10X SOLUTION, 1 LITER
SDP
Catalog No. 50489080
Encompass_Preferred
TBE is a common buffer used in gel electrophoresis.
Supplied as a 1X concentrated solution containing:
89mM Tris Base
89mM Boric Acid
2mM EDTA disodium salt
pH (1X solution @ 25°:C): 8.3+0.1
Catalog No. 50-489-080 Supplier Research Products International Corp Supplier No. T320201000.0
TBE is a common buffer used in gel electrophoresis.
Supplied as a 1X concentrated solution containing:
89mM Tris Base
89mM Boric Acid
2mM EDTA disodium salt
pH (1X solution @ 25°:C): 8.3+0.1