Learn More
Research Products International Corp RPI TBE 10X Solution [TRIS-borate-EDTA 10X Solution], 10 Liters
SDP
Catalog No. 501977326
Encompass_Preferred
TBE is a common buffer used in gel electrophoresis.
Supplied as a 1X concentrated solution containing:
89mM Tris Base
89mM Boric Acid
2mM EDTA disodium salt
pH (1X solution @ 25°:C): 8.3+0.1
Catalog No. 50-197-7326 Supplier Research Products International Corp Supplier No. T3202010000.0
TBE is a common buffer used in gel electrophoresis.
Supplied as a 1X concentrated solution containing:
89mM Tris Base
89mM Boric Acid
2mM EDTA disodium salt
pH (1X solution @ 25°:C): 8.3+0.1