Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals TBE (10X), TRIS + borate + EDTA

Catalog No. AAJ62449K7
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62449K7 4 L
AAJ62449K2 1 L
AAJ62449K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62449K7 Supplier Thermo Scientific Chemicals Supplier No. J62449K7
  • $233.00
    $175.65 / Each of 1

    Save $57.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. TBE Buffer is used for polyacrylamide and agarose gel electrophoresis. It is ideally suited for DNA and RNA separation on longer run, higher voltage or amperage gels. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel.5 TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids. Tris-Borate-EDTA buffer has been used for pulsed-field gel electrophoresis (PFGE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer
Description Colorless
Concentration 10X
Physical Form Liquid
Quantity 4 L
My Thermo Scientific brand TBE buffer has precipitated out of solution. What can I do?

Concentrations of 5X Tris Borate EDTA (TBE) buffer or greater may precipitate due to the nature of the product. This precipitation is a result of nucleation of salt crystals by dust particles or other insoluble materials. In some cases, we recommend heating the buffer to ~ 50°C to re-dissolve the TBE into solution. Alternately, to avoid precipitation of high concentration TBE buffers, after the solution is prepared, you can filter it through a 0.2 µm cellulose acetate or cellulose nitrate filter. These treatments have no deleterious effect on the buffering properties of TBE.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.