Learn More
Trajan Scientific TBE 5VSD-150/220 5MTRSTBE 5VS
SDP
Catalog No. 502530224
Encompass_Preferred
Trajan Deactivated (Methyl) Fused Silica Tubing, Diameter Inner: 0.005 in., 0.15 mm, Diameter Outer: 0.008 in., 0.22 mm, Overall Length: 16.4 ft., 5 m, Capillary Tubing, For Use With: Ideal for Biotechnology Applications, and Organic and Aqueous Solvents
Catalog No. 50-253-0224 Supplier Trajan Scientific Supplier No. 0624461
Trajan Deactivated (Methyl) Fused Silica Tubing, Diameter Inner: 0.005 in., 0.15 mm, Diameter Outer: 0.008 in., 0.22 mm, Overall Length: 16.4 ft., 5 m, Capillary Tubing, For Use With: Ideal for Biotechnology Applications, and Organic and Aqueous Solvents