Learn More
Crescent Chemical Co Inc TBE BUFFER (10X)
SDP
Catalog No. 50248613
Encompass_Preferred
TBE Buffer (10x) 10 x concentrated aqueous solution.TBE Buffer is widely used for the electrophoresis of nucleic acids and has a higher buffer capacity than TAE. It can be used for DNA and RNA polyacrylamide and agarose gel electrophoresis. Size - 2x500ML
Catalog No. 50-248-613 Supplier Crescent Chemical Co Inc Supplier No. SE42557.01
Add to Cart
Add to Cart
TBE Buffer (10x) 10 x concentrated aqueous solution.TBE Buffer is widely used for the electrophoresis of nucleic acids and has a higher buffer capacity than TAE. It can be used for DNA and RNA polyacrylamide and agarose gel electrophoresis. Size - 2x500ML