Learn More
Research Products International Corp TBE Buffer [TRIS-Borate-EDTA], 10X Powder, 1702.9 Grams of Powder Makes 10 Liters of Solution
SDP
Catalog No. 50213689
Encompass_Preferred
Dry homogeneous blend of molecular biology grade TRIS base, Boric Acid and EDTA.
For use in gel electrophoresis.
Catalog No. 50-213-689 Supplier Research Products International Corp Supplier No. T2202010.0
Dry homogeneous blend of molecular biology grade TRIS base, Boric Acid and EDTA.
For use in gel electrophoresis.