Learn More
Bioworld TBE Buffer (25X), 4 L
Catalog No. 50196694
Encompass_Preferred
Product DescriptionTBE (Tris-Borate-EDTA) has a liquid concentrate of 25X, where 1X contains 0.089M Tris, 0.089M Borate, and 0.002M EDTA. 1L of concentrate makes 25L of 1X buffer. Tris solutions are effective buffers for slightly basic conditions. Borate, a derivative of boric acid, is a weak proton donor but a Lewis acid that can accept an electron pair. EDTA is a chelator of divalent cations, mainly Mg2+. TBE is most commonly used for nucleic acid electrophoresis.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-196-694 Supplier Bioworld Supplier No. 105300222
Product DescriptionTBE (Tris-Borate-EDTA) has a liquid concentrate of 25X, where 1X contains 0.089M Tris, 0.089M Borate, and 0.002M EDTA. 1L of concentrate makes 25L of 1X buffer. Tris solutions are effective buffers for slightly basic conditions. Borate, a derivative of boric acid, is a weak proton donor but a Lewis acid that can accept an electron pair. EDTA is a chelator of divalent cations, mainly Mg2+. TBE is most commonly used for nucleic acid electrophoresis.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.