Learn More
Biotium TBE Buffer, 5X
Catalog No. 501964671
Encompass_Preferred
5X TBE (Tris-Borate-EDTA) is a concentrated buffer solution in deionized water. TBE is used for polyacrylamide and agarose gel electrophoresis. 1X solution: 89 mM Tris,89 mM boric acid, 2 mM EDTA. Store at room temperature.
Catalog No. 50-196-4671 Supplier Biotium Supplier No. 41006
5X TBE (Tris-Borate-EDTA) is a concentrated buffer solution in deionized water. TBE is used for polyacrylamide and agarose gel electrophoresis. 1X solution: 89 mM Tris,89 mM boric acid, 2 mM EDTA. Store at room temperature.