Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBE Buffer Mix (Dry Powder Mix of Tris/Boric Acid/EDTA/Electrophoresis), Fisher BioReagents

Catalog No. BP139686 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
86.1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1396-86 86.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP1396-86

missing translation for 'validMainframeMsg'

Electrophoresis buffer.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBE Buffer Mix
CAS 77-86-1, 6381-92-6, 10043-35-3
Color White
pH 8.2 to 8.4
Physical Form Solid
Quantity 86.1 g
DNase DNase free
Packaging Amber glass bottle
Synonym Tris-Boric Acid-EDTA Buffer Mix
Grade Electrophoresis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.