Learn More
Labcon North America 15mL UVSafe Centrifuge Tubes with Plug Style Caps, 50 per Bag, Sterile
Catalog No. NC1587906
Encompass_Preferred
Tube, Conical centrifuge; Labcon; UVSafe; 15mL; Sterile; With attached plug style caps; Block 100% of UVA, UVB and UVC light but passes visible light; white printed graduations; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 12,500 x G; Freezable -80C; Autoclavable tubes; Capacity: 15mL; Teal color caps; 50 tubes per bag/10 bags per case; 17mm W. 118mm H; Made in USA
Catalog No. NC1587906 Supplier Labcon North America Supplier No. 31324450089
Tube, Conical centrifuge; Labcon; UVSafe; 15mL; Sterile; With attached plug style caps; Block 100% of UVA, UVB and UVC light but passes visible light; white printed graduations; ATP, RNase/DNase and Endotoxin Free (non-pyrogenic); Validated Sterilization (ISO 11137) for use in Research and Medical labs; Polypropylene USP Class VI resin; Centrifuging to 12,500 x G; Freezable -80C; Autoclavable tubes; Capacity: 15mL; Teal color caps; 50 tubes per bag/10 bags per case; 17mm W. 118mm H; Made in USA