Learn More
Bio Rad Laboratories TBE MINIGELS 12WELL W/ETBR 1%
Catalog No. NC9830307
Encompass
Gel, Mini, ReadyAgarose Precast TBE, 1.0% plus Ethidium Bromide, 12-well.
Catalog No. NC9830307 Supplier Bio Rad Laboratories Supplier No. 1613010
Gel, Mini, ReadyAgarose Precast TBE, 1.0% plus Ethidium Bromide, 12-well.