Learn More
Bioworld TBE Sample Buffer (4X), 50 mL
Supplier:  Bioworld 105700272
Encompass_Preferred
Tris-Borate Buffer, EDTA (TBE) Sample buffer for nucleic acid analysis. Technical Information: 4X concentration Tris base (270mM) Boric Acid (270mM) Glycerol (36%) Bromophenol blue G (0.025%) Xylene Cyanol (0.025%) EDTA (6mM)
Catalog No. 50-196-794