Learn More
Bioworld TBE Sample Buffer (6X), 50 mL
Catalog No. 50196796
Encompass_Preferred
Product DescriptionTBE (Tris-Borate-EDTA) is a sample buffer used for nucleic acid analysis. It has a 6X concentration, 270 mM Tris base, 270 mM boric acid, 40% glycerol, 0.025% bromophenol blue G, 0.025% xylene cyanol, and 6 mM of EDTA.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-196-796 Supplier Bioworld Supplier No. 105700282
Product DescriptionTBE (Tris-Borate-EDTA) is a sample buffer used for nucleic acid analysis. It has a 6X concentration, 270 mM Tris base, 270 mM boric acid, 40% glycerol, 0.025% bromophenol blue G, 0.025% xylene cyanol, and 6 mM of EDTA.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.