Learn More
Kd Medical Inc TBE (TRIS-BORATE EDTA), 10X
SDP
Catalog No. 501031381
Encompass_Preferred
Tris Borate EDTA is used for polyacrylamide and agarose gel electrophoresis. 10X TBE is concentrated and ready to be diluted to a 1X working solution. One 500ml bottle.
Catalog No. 50-103-1381 Supplier Kd Medical Inc Supplier No. RGE3330
Tris Borate EDTA is used for polyacrylamide and agarose gel electrophoresis. 10X TBE is concentrated and ready to be diluted to a 1X working solution. One 500ml bottle.