Learn More
Bioworld TBE Urea Running Buffer (10X), 4 L
Catalog No. 50196697
Encompass_Preferred
Product DescriptionTBE(Tris-Borate-EDTA) urea running buffer is used for protein analysis. It has a 10X concentration and contains 0.89M Tris base, 0.89M boric acid, 20 mM EDTA, and a pH at 8.3.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-196-697 Supplier Bioworld Supplier No. 105300243
Product DescriptionTBE(Tris-Borate-EDTA) urea running buffer is used for protein analysis. It has a 10X concentration and contains 0.89M Tris base, 0.89M boric acid, 20 mM EDTA, and a pH at 8.3.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.