Learn More
Bioworld TBE Urea Sample Buffer (2X), 25 mL
Catalog No. 50196698
Encompass_Preferred
Product DescriptionTBE(Tris-Borate-EDTA) urea sample buffer is used for nucleic acid analysis. It has a 2X concentration and contains 45 mM Tris base, 45 mM Boric acid, 6% Ficol, 3.5M Urea, 0.005% Bromophenol blue, 0.025% Xylene and 1 mM EDTA.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-196-698 Supplier Bioworld Supplier No. 105300251
Product DescriptionTBE(Tris-Borate-EDTA) urea sample buffer is used for nucleic acid analysis. It has a 2X concentration and contains 45 mM Tris base, 45 mM Boric acid, 6% Ficol, 3.5M Urea, 0.005% Bromophenol blue, 0.025% Xylene and 1 mM EDTA.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.