Learn More
Beckman Coulter Beckman Coulter 400 μL Polyethylene Microcentrifuge Tube, Heparin-Lithium-Fluoride Coated
Catalog No. NC2946670
Encompass_Preferred
Beckman Coulter 400 μL polyethylene microcentrifuge tube coated with heparin-lithium-fluoride for anticoagulant sample collection and processing. Intended for use with fixed-angle rotors and compatible 0.25/0.40 mL adapters; suitable for routine clinical and research centrifugation workflows.
  • 400 μL nominal volume.
  • Polyethylene material.
  • Heparin-lithium-fluoride coating.
  • Size: 7 x 40 mm.
  • Pack quantity: 1,000 per pack.
  • Designed for use with fixed-angle rotors and 0.25/0.40 mL adapters.
  • Maximum recommended speed: 11,500 rpm.
Catalog No. NC2946670 Supplier Beckman Coulter Supplier No. 652824
Beckman Coulter 400 μL polyethylene microcentrifuge tube coated with heparin-lithium-fluoride for anticoagulant sample collection and processing. Intended for use with fixed-angle rotors and compatible 0.25/0.40 mL adapters; suitable for routine clinical and research centrifugation workflows.
  • 400 μL nominal volume.
  • Polyethylene material.
  • Heparin-lithium-fluoride coating.
  • Size: 7 x 40 mm.
  • Pack quantity: 1,000 per pack.
  • Designed for use with fixed-angle rotors and 0.25/0.40 mL adapters.
  • Maximum recommended speed: 11,500 rpm.