Learn More
Enzo Life Sciences Tick-borne Encephalitis Virus Envelope protein, (recombinant) (mouse Fc-tag) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2790100
Encompass_Preferred
Recombinant Tick-borne encephalitis virus (TBEV) protein produced in HEK293 cells, incorporating a C-terminal mouse Fc-tag. Recombinant Tick-borne encephalitis virus (TBEV) protein produced in HEK293 cells, incorporating a C-terminal mouse Fc-tag. Alternative name: TBEV Envelope protein. Formulation: Liquid. In DPBS, pH 7.4. Contains traces of CHAPS. Source: Produced in HEK293 cells. Tick-borne encephalitis virus envelope protein is fused at the C-terminus to a mouse Fc-tag. Long Term Storage: -80°C. Applications: ELISA Vaccine research, LINE assay, Antibody capture.
Catalog No. 50-201-4091