Learn More
ACROBiosystems TBEV European subtype strain
Supplier:  ACROBiosystems ENET52H450UG
Encompass_Preferred
TBEV (European subtype strain Neudoerfl) Envelope protein E His Tag (ENE-T52H4) is expressed from human 293 cells (HEK293). It contains AA Thr 118 - Lys 675 (Accession # P14336).
Catalog No. 50-253-5107