Learn More
Creative Biomart Recombinant Tick-Borne Encephalitis Virus gE Antigen
Catalog No. 502084447
Encompass_Preferred
The E. coli derived recombinant protein contains the Tick-borne Encephalitis Virus glycoprotein E middle regions, 50-250 amino acids.
Catalog No. 50-208-4447 Supplier Creative Biomart Supplier No. TBE069
The E. coli derived recombinant protein contains the Tick-borne Encephalitis Virus glycoprotein E middle regions, 50-250 amino acids.